Methyl (R)-2-amino-3-(4-bromophenyl)propanoate 98%

Lieferant: Apollo Scientific

OR471715-1G OR471715-250MG
APOSOR471715-1GEA 679 EUR
APOSOR471715-1G APOSOR471715-250MG
Methyl (R)-2-amino-3-(4-bromophenyl)propanoate 98%
Methyl (R)-2-amino-3-(4-bromophenyl)propanoate
Formel: C₁₀H₁₂BrNO₂
Molecular Weight: 258,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08058268
CAS-Nummer: 122332-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR