Methyl 2-(6-bromo-3-pyridyl)acetate 95%

Lieferant: Apollo Scientific

OR471726-1G OR471726-250MG
APOSOR471726-1GEA 513 EUR
APOSOR471726-1G APOSOR471726-250MG
Methyl 2-(6-bromo-3-pyridyl)acetate 95%
Methyl 2-(6-bromo-3-pyridyl)acetate
Formel: C₈H₈BrNO₂
Molecular Weight: 230.059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18256344
CAS-Nummer: 1256824-78-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR