6-Cyclopropylpyridine-3-methanamine 97%

Lieferant: Apollo Scientific

OR471727-100MG OR471727-1G OR471727-250MG
APOSOR471727-100MGEA 551 EUR
APOSOR471727-100MG APOSOR471727-1G APOSOR471727-250MG
6-Cyclopropylpyridine-3-methanamine 97%
6-Cyclopropylpyridine-3-methanamine
Formel: C₉H₁₂N₂
Molecular Weight: 148,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18255577
CAS-Nummer: 1256823-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR