4-(1H-1,2,3-Triazol-1-yl)phenylboronic acid pinacol ester 97%
Lieferant: Apollo Scientific
Synonyme:
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-1,2,3-triazole
OR471729-100MG
OR471729-1G
OR471729-250MG
APOSOR471729-100MGEA
168
EUR
APOSOR471729-100MG
APOSOR471729-1G
APOSOR471729-250MG
4-(1H-1,2,3-Triazol-1-yl)phenylboronic acid pinacol ester 97%
4-(1H-1,2,3-Triazol-1-yl)phenylboronic acid pinacol ester
Formel:
C₁₄H₁₈BN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22570712 CAS-Nummer: 1101174-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...