1-Methylimidazole-5-carboxamide 95+%

Lieferant: Apollo Scientific

OR4728-1G OR4728-10G OR4728-5G
APOSOR4728-1GEA 243 EUR
APOSOR4728-1G APOSOR4728-10G APOSOR4728-5G
1-Methylimidazole-5-carboxamide 95+%
1-Methylimidazole-5-carboxamide
Formel: C₅H₇N₃O
Molecular Weight: 125,129 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01567302
CAS-Nummer: 40637-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR