tert-Butyl [(3S)-pyrrolidin-3-ylmethyl]carbamate hydrochloride
Lieferant: Apollo Scientific
Synonyme:
(S)-tert-Butyl (pyrrolidin-3-ylmethyl)carbamate hydrochloride
OR47363-1G
OR47363-250MG
APOSOR47363-1GEA
128
EUR
APOSOR47363-1G
APOSOR47363-250MG
tert-Butyl [(3S)-pyrrolidin-3-ylmethyl]carbamate hydrochloride
tert-Butyl [(3S)-pyrrolidin-3-ylmethyl]carbamate hydrochloride
Formel:
C₁₀H₂₀N₂O₂·ClH Molecular Weight: 236,739 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11101394 CAS-Nummer: 1075260-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...