N-[1-(4-Bromophenyl)ethyl]cyclopropanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR475075-1G OR475075-250MG
APOSOR475075-1GEA 133 EUR
APOSOR475075-1G APOSOR475075-250MG
N-[1-(4-Bromophenyl)ethyl]cyclopropanamine hydrochloride
N-[1-(4-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Formel: C₁₁H₁₄BrN·ClH
Molecular Weight: 276,601 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31695688
CAS-Nummer: 2367002-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR