N-[(1R)-1-(3-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR475182-1G
OR475182-250MG
APOSOR475182-1GEA
201
EUR
APOSOR475182-1G
APOSOR475182-250MG
N-[(1R)-1-(3-Bromophenyl)ethyl]cyclopropanamine hydrochloride
N-[(1R)-1-(3-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Formel:
C₁₁H₁₄BrN·ClH Molecular Weight: 276,601 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD32066849 CAS-Nummer: 2414145-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...