(4-Bromo-1-cyclopropyl-1H-pyrazol-3-yl)methanol

Lieferant: Apollo Scientific

OR475189-1G OR475189-250MG
APOSOR475189-1GEA 654 EUR
APOSOR475189-1G APOSOR475189-250MG
(4-Bromo-1-cyclopropyl-1H-pyrazol-3-yl)methanol
(4-Bromo-1-cyclopropyl-1H-pyrazol-3-yl)methanol
Formel: C₇H₉BrN₂O
Molecular Weight: 217,063 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32200421
CAS-Nummer: 2413937-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR