5-(1H-Pyrrol-1-yl)nicotinic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(1H-Pyrrol-1-yl)pyridine-3-carboxylic acid

OR4766-250MG
APOSOR4766-250MGEA 105 EUR
APOSOR4766-250MG
5-(1H-Pyrrol-1-yl)nicotinic acid 95%
5-(1H-Pyrrol-1-yl)nicotinic acid
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04971938
CAS-Nummer: 690632-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR