5-Aminobenzo[b]thiophene

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Aminobenzothiophene, 1-Benzothiophen-5-amine, 5-Amino-1-benzothiophene

OR47828-1G OR47828-250MG OR47828-5G
APOSOR47828-1GEA 78.3 EUR
APOSOR47828-1G APOSOR47828-250MG APOSOR47828-5G
5-Aminobenzo[b]thiophene
5-Aminobenzo[b]thiophene
Formel: C₈H₇NS
Molecular Weight: 149.213 g/mol
Schmelzpunkt: 72…75 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115381
CAS-Nummer: 20532-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR