Ethyl 3-amino-5-bromopyridine-2-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-5-bromo-2-(ethoxycarbonyl)pyridine, Ethyl 3-amino-5-bromopicolinate
OR47833-250MG
OR47833-1G
APOSOR47833-250MGEA
149
EUR
APOSOR47833-250MG
APOSOR47833-1G
Ethyl 3-amino-5-bromopyridine-2-carboxylate 95%
Ethyl 3-amino-5-bromopyridine-2-carboxylate
Formel:
C₈H₉BrN₂O₂ Molecular Weight: 245.073 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10566234 CAS-Nummer: 1334405-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...