2-Bromo-5-cyclopropylaniline
Lieferant: Apollo Scientific
|
Achtung
|
OR47841-1G
OR47841-250MG
OR47841-5G
APOSOR47841-1GEA
575
EUR
APOSOR47841-1G
APOSOR47841-250MG
APOSOR47841-5G
2-Bromo-5-cyclopropylaniline
2-Bromo-5-cyclopropylaniline
Formel:
C₉H₁₀BrN Lagertemperatur: Gefrierschrank |
MDL:
MFCD21099977 CAS-Nummer: 1353855-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...