3-Bromo-4-cyclopropylaniline

Lieferant: Apollo Scientific
Achtung

OR47843-1G OR47843-250MG OR47843-5G
APOSOR47843-1GEA 575 EUR
APOSOR47843-1G APOSOR47843-250MG APOSOR47843-5G
3-Bromo-4-cyclopropylaniline
3-Bromo-4-cyclopropylaniline
Formel: C₉H₁₀BrN
Molecular Weight: 212,086 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD21100011
CAS-Nummer: 1353854-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR