1-Bromo-3-(tert-butoxy)benzene
Lieferant: Apollo Scientific
OR47857-1G
OR47857-250MG
APOSOR47857-1GEA
255
EUR
APOSOR47857-1G
APOSOR47857-250MG
1-Bromo-3-(tert-butoxy)benzene
1-Bromo-3-(tert-butoxy)benzene
Formel:
C₁₀H₁₃BrO Molecular Weight: 229.114 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18917788 CAS-Nummer: 99376-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...