2-Bromo-5-chloroisonicotinaldehyde 97%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-5-chloropyridine-4-carboxaldehyde
OR47896-1G
OR47896-250MG
OR47896-5G
APOSOR47896-1GEA
190
EUR
APOSOR47896-1G
APOSOR47896-250MG
APOSOR47896-5G
2-Bromo-5-chloroisonicotinaldehyde 97%
2-Bromo-5-chloroisonicotinaldehyde
Formel:
C₆H₃BrClNO Molecular Weight: 220,451 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09907898 CAS-Nummer: 921630-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...