3-Chloro-5,6-dimethylpyridazine-4-carbonitrile

Lieferant: Apollo Scientific

OR47921-100MG OR47921-1G OR47921-250MG
APOSOR47921-100MGEA 108 EUR
APOSOR47921-100MG APOSOR47921-1G APOSOR47921-250MG
3-Chloro-5,6-dimethylpyridazine-4-carbonitrile
3-Chloro-5,6-dimethylpyridazine-4-carbonitrile
Formel: C₇H₆ClN₃
Molecular Weight: 167.596 g/mol
Schmelzpunkt: 79…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09802146
CAS-Nummer: 93824-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR