Ethyl 5-amino-4-bromo-1-(3-pyridyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480067-1G
APOSOR480067-1GEA 1340 EUR
APOSOR480067-1G
Ethyl 5-amino-4-bromo-1-(3-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(3-pyridyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₁BrN₄O₂
Molecular Weight: 311.135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135160
CAS-Nummer: 1427013-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR