(2R)-2-Amino-3-methylsulphinyl-propanoic acid

Lieferant: Apollo Scientific

Synonyme: (2R)-2-Amino-3-(methylsulphinyl)propanoic acid

OR480202-1G OR480202-500MG OR480202-5G
APOSOR480202-1GEA 245 EUR
APOSOR480202-1G APOSOR480202-500MG APOSOR480202-5G
(2R)-2-Amino-3-methylsulphinyl-propanoic acid
(2R)-2-Amino-3-methylsulphinyl-propanoic acid
Formel: C₄H₉NO₃S
Molecular Weight: 151,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00152132
CAS-Nummer: 6853-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR