1,2,2,6,6-Pentamethyl-4-piperidone 97%

Lieferant: Apollo Scientific

Synonyme: 1,2,2,6,6-Pentamethylpiperidin-4-one

OR480208-1G OR480208-25G OR480208-5G
APOSOR480208-1GEA 222 EUR
APOSOR480208-1G APOSOR480208-25G APOSOR480208-5G
1,2,2,6,6-Pentamethyl-4-piperidone 97%
1,2,2,6,6-Pentamethyl-4-piperidone
Formel: C₁₀H₁₉NO
Molecular Weight: 169,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234684
CAS-Nummer: 5554-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR