Methyl 5-amino-2,4-dimethyl-benzoate

Lieferant: Apollo Scientific

OR480210-1G OR480210-250MG OR480210-500MG
APOSOR480210-1GEA 306 EUR
APOSOR480210-1G APOSOR480210-250MG APOSOR480210-500MG
Methyl 5-amino-2,4-dimethyl-benzoate
Methyl 5-amino-2,4-dimethyl-benzoate
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00230534
CAS-Nummer: 140112-97-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR