(2R,4S)-4-Hydroxypiperidine-2-carboxylic acid 95%
Lieferant: Apollo Scientific
OR480267-1G
OR480267-250MG
OR480267-500MG
APOSOR480267-1GEA
864
EUR
APOSOR480267-1G
APOSOR480267-250MG
APOSOR480267-500MG
(2R,4S)-4-Hydroxypiperidine-2-carboxylic acid 95%
(2R,4S)-4-Hydroxypiperidine-2-carboxylic acid
Formel:
C₆H₁₁NO₃·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110198 CAS-Nummer: 175671-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...