(2-Chloro-6-methyl-3-pyridyl)methanol 97%

Lieferant: Apollo Scientific

OR480273-10G OR480273-1G OR480273-5G
APOSOR480273-10GEA 1130 EUR
APOSOR480273-10G APOSOR480273-1G APOSOR480273-5G
(2-Chloro-6-methyl-3-pyridyl)methanol 97%
(2-Chloro-6-methyl-3-pyridyl)methanol
Formel: C₇H₈ClNO
Molecular Weight: 157,598 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847244
CAS-Nummer: 689259-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR