1-(Hydroxymethyl)cyclopropanecarboxamide 95%
Lieferant: Apollo Scientific
OR480277-10G
OR480277-1G
OR480277-5G
APOSOR480277-10GEA
1280
EUR
APOSOR480277-10G
APOSOR480277-1G
APOSOR480277-5G
1-(Hydroxymethyl)cyclopropanecarboxamide 95%
1-(Hydroxymethyl)cyclopropanecarboxamide
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12198439 CAS-Nummer: 1123169-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...