4-Bromo-5-methoxy-1H-indole 95%

Lieferant: Apollo Scientific

OR480279-1G OR480279-250MG OR480279-500MG
APOSOR480279-1GEA 235 EUR
APOSOR480279-1G APOSOR480279-250MG APOSOR480279-500MG
4-Bromo-5-methoxy-1H-indole 95%
4-Bromo-5-methoxy-1H-indole
Formel: C₉H₈BrNO
Molecular Weight: 226.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12973810
CAS-Nummer: 90858-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR