5-(Methoxymethyl)-1-methyl-pyrazol-3-amine 95%

Lieferant: Apollo Scientific

OR480282-1G OR480282-250MG OR480282-500MG
APOSOR480282-1GEA 698 EUR
APOSOR480282-1G APOSOR480282-250MG APOSOR480282-500MG
5-(Methoxymethyl)-1-methyl-pyrazol-3-amine 95%
5-(Methoxymethyl)-1-methyl-pyrazol-3-amine
Formel: C₆H₁₁N₃O
Molecular Weight: 141.171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20491525
CAS-Nummer: 1328640-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR