2-Piperazin-1-yl-1,3-benzoxazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Piperazin-1-yl)benzo[d]oxazole
OR480292-10G
OR480292-1G
OR480292-5G
APOSOR480292-10GEA
1030
EUR
APOSOR480292-10G
APOSOR480292-1G
APOSOR480292-5G
2-Piperazin-1-yl-1,3-benzoxazole 95%
2-Piperazin-1-yl-1,3-benzoxazole
Formel:
C₁₁H₁₃N₃O Schmelzpunkt: 75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04035522 CAS-Nummer: 111628-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...