DL-Propargylglycine 97%
Lieferant: Apollo Scientific
Synonyme:
2-Aminopent-4-ynoic acid, PAG, 2-Amino-4-pentynoic acid, H-DL-Pra-OH
OR480425-1G
OR480425-5G
APOSOR480425-1GEA
52.2
EUR
APOSOR480425-1G
APOSOR480425-5G
DL-Propargylglycine 97%
DL-Propargylglycine
Formel:
C₅H₇NO₂ Molecular Weight: 113,116 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056728 CAS-Nummer: 50428-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...