2-Amino-2-(4-ethoxyphenyl)acetic acid

Lieferant: Apollo Scientific

OR480447-1G OR480447-5G
APOSOR480447-1GEA 114 EUR
APOSOR480447-1G APOSOR480447-5G
2-Amino-2-(4-ethoxyphenyl)acetic acid
2-Amino-2-(4-ethoxyphenyl)acetic acid
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665360
CAS-Nummer: 299168-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR