2-Amino-2-(4-isopropoxyphenyl)acetic acid

Lieferant: Apollo Scientific

OR480455-1G OR480455-2G OR480455-5G
APOSOR480455-1GEA 171 EUR
APOSOR480455-1G APOSOR480455-2G APOSOR480455-5G
2-Amino-2-(4-isopropoxyphenyl)acetic acid
2-Amino-2-(4-isopropoxyphenyl)acetic acid
Formel: C₁₁H₁₅NO₃
Molecular Weight: 209,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06653255
CAS-Nummer: 318270-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR