Amino(5-bromo-2,4-dimethoxyphenyl)acetic acid
Lieferant: Apollo Scientific
OR480465-1G
OR480465-2G
OR480465-5G
APOSOR480465-1GEA
306
EUR
APOSOR480465-1G
APOSOR480465-2G
APOSOR480465-5G
Amino(5-bromo-2,4-dimethoxyphenyl)acetic acid
Amino(5-bromo-2,4-dimethoxyphenyl)acetic acid
Formel:
C₁₀H₁₂BrNO₄ Molecular Weight: 290.111 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02662702 CAS-Nummer: 500718-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...