2-Amino-2-(4-benzyloxyphenyl)acetic acid

Lieferant: Apollo Scientific

OR480466-2G OR480466-5G
APOSOR480466-2GEA 260 EUR
APOSOR480466-2G APOSOR480466-5G
2-Amino-2-(4-benzyloxyphenyl)acetic acid
2-Amino-2-(4-benzyloxyphenyl)acetic acid
Formel: C₁₅H₁₅NO₃
Molecular Weight: 257.285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02662410
CAS-Nummer: 72028-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR