2-Benzothiazole-2-oxyacetic acid 97%

Lieferant: Apollo Scientific

OR480481-1G OR480481-5G
APOSOR480481-1GEA 159 EUR
APOSOR480481-1G APOSOR480481-5G
2-Benzothiazole-2-oxyacetic acid 97%
2-Benzothiazole-2-oxyacetic acid
Formel: C₉H₇NO₃S
Molecular Weight: 209,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368969
CAS-Nummer: 2875-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR