5-Bromo-3-chloropyridazine 90%
Lieferant: Apollo Scientific
|
Achtung
|
OR480780-1G
OR480780-250MG
OR480780-500MG
OR480780-5G
APOSOR480780-1GEA
161
EUR
APOSOR480780-1G
APOSOR480780-250MG
APOSOR480780-500MG
APOSOR480780-5G
5-Bromo-3-chloropyridazine 90%
5-Bromo-3-chloropyridazine
Formel:
C₄H₂BrClN₂ Molecular Weight: 193,429 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13190293 CAS-Nummer: 1196155-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...