5-Bromo-2-cyclopropylaniline

Lieferant: Apollo Scientific

OR48081-1G OR48081-250MG OR48081-5G
APOSOR48081-1GEA 575 EUR
APOSOR48081-1G APOSOR48081-250MG APOSOR48081-5G
5-Bromo-2-cyclopropylaniline
5-Bromo-2-cyclopropylaniline
Formel: C₉H₁₀BrN
Molecular Weight: 212,086 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD10465684
CAS-Nummer: 104902-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR