4-[[(1S)-2-Hydroxy-1-[(4-hydroxyphenyl)methyl]-2-oxo-ethyl]amino]-4-oxo-butanoic acid 90%

Lieferant: Apollo Scientific

OR480811-1G OR480811-5G
APOSOR480811-1GEA 482 EUR
APOSOR480811-1G APOSOR480811-5G
4-[[(1S)-2-Hydroxy-1-[(4-hydroxyphenyl)methyl]-2-oxo-ethyl]amino]-4-oxo-butanoic acid 90%
4-[[(1S)-2-Hydroxy-1-[(4-hydroxyphenyl)methyl]-2-oxo-ethyl]amino]-4-oxo-butanoic acid
Formel: C₁₃H₁₅NO₆
Molecular Weight: 281,261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18252486
CAS-Nummer: 374816-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR