(1R,3S)-3-Aminocyclohexanecarboxylic acid hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR480822-1G OR480822-5G
APOSOR480822-1GEA 215 EUR
APOSOR480822-1G APOSOR480822-5G
(1R,3S)-3-Aminocyclohexanecarboxylic acid hydrochloride 95%
(1R,3S)-3-Aminocyclohexanecarboxylic acid hydrochloride
Formel: C₇H₁₃NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28166349
CAS-Nummer: 2058044-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR