5-Bromo-3-hydroxy-3H-isobenzofuran-1-one 90%, tech
Lieferant: Apollo Scientific
OR480842-1G
OR480842-5G
OR480842-250MG
APOSOR480842-1GEA
116
EUR
APOSOR480842-1G
APOSOR480842-5G
APOSOR480842-250MG
5-Bromo-3-hydroxy-3H-isobenzofuran-1-one 90%, tech
5-Bromo-3-hydroxy-3H-isobenzofuran-1-one
Formel:
C₈H₅BrO₃ Molecular Weight: 229,028 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09836746 CAS-Nummer: 102126-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...