8-Boc-3-amino-1-oxa-8-azaspiro[4.5]decane
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 3-amino-1-oxa-8-azaspiro[4.5]decane-8-carboxylate
OR48085-1G
OR48085-250MG
APOSOR48085-1GEA
470
EUR
APOSOR48085-1G
APOSOR48085-250MG
8-Boc-3-amino-1-oxa-8-azaspiro[4.5]decane
8-Boc-3-amino-1-oxa-8-azaspiro[4.5]decane
Formel:
C₁₃H₂₄N₂O₃ Molecular Weight: 256,341 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12198529 CAS-Nummer: 1160246-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...