3-(3-Hydroxyphenyl)-piperazine-1-carboxylic acid tert-butyl ester 95%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 3-(3-hydroxyphenyl)piperazine-1-carboxylate
OR480858-1G
OR480858-250MG
APOSOR480858-1GEA
1500
EUR
APOSOR480858-1G
APOSOR480858-250MG
3-(3-Hydroxyphenyl)-piperazine-1-carboxylic acid tert-butyl ester 95%
3-(3-Hydroxyphenyl)-piperazine-1-carboxylic acid tert-butyl ester
Formel:
C₁₅H₂₂N₂O₃ Molecular Weight: 278.347 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07021282 CAS-Nummer: 889956-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...