2-Bromo-4-iodo-1-isopropoxybenzene 95%

Lieferant: Apollo Scientific

OR48087-1G OR48087-250MG
APOSOR48087-1GEA 654 EUR
APOSOR48087-1G APOSOR48087-250MG
2-Bromo-4-iodo-1-isopropoxybenzene 95%
2-Bromo-4-iodo-1-isopropoxybenzene
Formel: C₉H₁₀BrIO
Molecular Weight: 340,984 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28129290
CAS-Nummer: 1369903-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR