6-(Chloromethyl)quinoline 95%
Lieferant: Apollo Scientific
OR480883-1G
OR480883-5G
APOSOR480883-1GEA
339
EUR
APOSOR480883-1G
APOSOR480883-5G
6-(Chloromethyl)quinoline 95%
6-(Chloromethyl)quinoline
Formel:
C₁₀H₈ClN Molecular Weight: 177,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09040306 CAS-Nummer: 2644-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...