1-tert-Butoxyvinyloxy-tert-butyldimethylsilane 90%
Lieferant: Apollo Scientific
OR480884-1G
OR480884-5G
APOSOR480884-1GEA
180
EUR
APOSOR480884-1G
APOSOR480884-5G
1-tert-Butoxyvinyloxy-tert-butyldimethylsilane 90%
1-tert-Butoxyvinyloxy-tert-butyldimethylsilane
Formel:
C₁₂H₂₆O₂Si Molecular Weight: 230.419 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12923224 CAS-Nummer: 74786-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...