2,7-Dichloroquinazoline 90%

Lieferant: Apollo Scientific

OR480887-1G
APOSOR480887-1GEA 1410 EUR
APOSOR480887-1G
2,7-Dichloroquinazoline 90%
2,7-Dichloroquinazoline
Formel: C₈H₄Cl₂N₂
Molecular Weight: 199,037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15527158
CAS-Nummer: 67092-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR