Ethyl 3-chloro-1H-indazole-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR48091-1G
OR48091-250MG
APOSOR48091-1GEA
27.7
EUR
APOSOR48091-1G
APOSOR48091-250MG
Ethyl 3-chloro-1H-indazole-5-carboxylate
Ethyl 3-chloro-1H-indazole-5-carboxylate
Formel:
C₁₀H₉ClN₂O₂ Molecular Weight: 224,644 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31699065 CAS-Nummer: 2121562-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...