Dimethyl 1-vinyl-1H-pyrazole-3,5-dicarboxylate

Lieferant: Apollo Scientific
Achtung

OR48095-1G OR48095-250MG
APOSOR48095-1GEA 30.6 EUR
APOSOR48095-1G APOSOR48095-250MG
Dimethyl 1-vinyl-1H-pyrazole-3,5-dicarboxylate
Dimethyl 1-vinyl-1H-pyrazole-3,5-dicarboxylate
Formel: C₉H₁₀N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD32111677
CAS-Nummer: 86797-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR