3-Cyclopropyl-2-thiophenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR48096-1G OR48096-250MG
APOSOR48096-1GEA 522 EUR
APOSOR48096-1G APOSOR48096-250MG
3-Cyclopropyl-2-thiophenesulphonyl chloride
3-Cyclopropyl-2-thiophenesulphonyl chloride
Formel: C₇H₇ClO₂S₂
Molecular Weight: 222,712 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31616404
CAS-Nummer: 2172096-17-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR