2-Bromo-6-chlorophenylacetylene 97%

Lieferant: Apollo Scientific
Achtung

OR48117-1G OR48117-250MG
APOSOR48117-1GEA 84.1 EUR
APOSOR48117-1G APOSOR48117-250MG
2-Bromo-6-chlorophenylacetylene 97%
2-Bromo-6-chlorophenylacetylene
Formel: C₈H₄BrCl
Molecular Weight: 215.474 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31700579
CAS-Nummer: 1856321-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR