2-(1-Aminocyclobutyl)ethan-1-ol

Lieferant: Apollo Scientific

OR48124-1G OR48124-500MG
APOSOR48124-1GEA 1150 EUR
APOSOR48124-1G APOSOR48124-500MG
2-(1-Aminocyclobutyl)ethan-1-ol
2-(1-Aminocyclobutyl)ethan-1-ol
Formel: C₆H₁₃NO
Molecular Weight: 115.174 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19215486
CAS-Nummer: 1132814-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR