(S)-5-(Aminomethyl)pyrrolidin-2-one 95%
Lieferant: Apollo Scientific
OR48125-1G
OR48125-250MG
APOSOR48125-1GEA
545
EUR
APOSOR48125-1G
APOSOR48125-250MG
(S)-5-(Aminomethyl)pyrrolidin-2-one 95%
(S)-5-(Aminomethyl)pyrrolidin-2-one
Formel:
C₅H₁₀N₂O Molecular Weight: 114,146 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06739064 CAS-Nummer: 145414-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...